Assay Details
| Gene Symbol: SPNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142448.1 | NP_001135920.1 | 13 | 1914 | 100 | |
| NM_001142449.1 | NP_001135921.1 | 10 | 1929 | 100 | ||
| NM_001142450.1 | NP_001135922.1 | 13 | 2064 | 100 | ||
| NM_001142451.1 | NP_001135923.1 | 11 | 1995 | 100 | ||
| NM_032038.2 | NP_114427.1 | 12 | 2151 | 100 | ||
| XM_006721096.3 | XP_006721159.1 | 12 | 1765 | 100 | ||
| XM_017023758.1 | XP_016879247.1 | 13 | 1933 | 100 | ||
| XM_017023759.1 | XP_016879248.1 | 13 | 1917 | 100 | ||
| GenBank mRNA | AF212371.1 | - | 12 | 2098 | 100 | |
| AF370423.1 | - | 13 | 2045 | 100 | ||
| AL390215.1 | - | 10 | 1891 | 100 | ||
| BC006156.1 | - | 12 | 1762 | 100 | ||
| BC008325.1 | - | 12 | 1976 | 3501826 | 100 | |
| BC038961.1 | - | 13 | 1928 | 6053095 | 100 | |
| BC047741.1 | - | 11 | 1969 | 5757339 | 100 | |
| BC065235.1 | - | 13 | 1896 | 6161778 | 100 | |
| CD368853.1 | - | 1 | 100 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SPNS1
Genomic Map