Assay Details
Gene Symbol: MICA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000247.2 | NP_000238.1 | 1 | 83 | 101 | |
GenBank mRNA | AK225393.1 | - | 1 | 65 | 101 | |
BC016929.1 | - | 1 | 71 | 4446575 | 101 | |
L14848.1 | - | 1 | 56 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06004073.hg.1, TC6_cox_hap2000234.hg.1, TC6_mann_hap4000215.hg.1, TC6_qbl_hap6000238.hg.1, TC6_ssto_hap7000188.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC016929.1,L14848.1,NM_000247.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
T cell mediated cytotoxicity
immune response to tumor cell
cellular response to DNA damage stimulus
response to heat
cytolysis
antigen processing and presentation
negative regulation of natural killer cell activation
natural killer cell mediated cytotoxicity
defense response to bacterium
negative regulation of natural killer cell mediated cytotoxicity
gamma-delta T cell activation
regulation of immune response
defense response to virus
antigen binding
protein binding
beta-2-microglobulin binding
natural killer cell lectin-like receptor binding
T cell mediated cytotoxicity
immune response to tumor cell
cellular response to DNA damage stimulus
response to heat
cytolysis
antigen processing and presentation
negative regulation of natural killer cell activation
natural killer cell mediated cytotoxicity
defense response to bacterium
negative regulation of natural killer cell mediated cytotoxicity
gamma-delta T cell activation
regulation of immune response
defense response to virus
antigen binding
protein binding
beta-2-microglobulin binding
natural killer cell lectin-like receptor binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06003455.hg.1 | TC06004073.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003455.hg.1 | ENST00000364810.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003455.hg.1 | ENST00000399172.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003455.hg.1 | ENST00000449934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003455.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003455.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000828.hg.1 | TC6_mann_hap4000215.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000828.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000828.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000854.hg.1 | TC6_cox_hap2000234.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000854.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000854.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000851.hg.1 | TC6_qbl_hap6000238.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000851.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000851.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003454.hg.1 | TC06004073.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003454.hg.1 | ENST00000364810.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003454.hg.1 | ENST00000399172.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003454.hg.1 | ENST00000449934.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003454.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003454.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000827.hg.1 | TC6_mann_hap4000215.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000827.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000827.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000850.hg.1 | TC6_qbl_hap6000238.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000850.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000850.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000646.hg.1 | TC6_ssto_hap7000188.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000646.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000646.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000855.hg.1 | TC6_cox_hap2000234.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000855.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000855.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000647.hg.1 | TC6_ssto_hap7000188.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000647.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000647.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198106.hg.1 | TC0600014101.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198105.hg.1 | TC0600014101.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000421350.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | ENST00000616296.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014101.hg.1 | NM_001289154.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | ENST00000449934.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | NM_000247.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | NM_001177519.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600198104.hg.1 | TC0600014101.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 109758 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 226440 | - | - |
