Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016139.hg.1 |
ENST00000300632.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016139.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016139.hg.1 |
TC11001274.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203306.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203306.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203306.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203306.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203306.hg.1 |
TC1100013130.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000300632.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
ENST00000356578.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NM_001007139.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013130.hg.1 |
NR_003512.3 |
Refseq |