Assay Details
| Gene Symbol: CCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002983.2 | NP_002974.1 | 1 | 43 | 95 | |
| GenBank mRNA | BX103455.1 | - | 1 | 43 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001385.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_002983.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
osteoblast differentiation
cell activation
monocyte chemotaxis
calcium ion transport
cellular calcium ion homeostasis
exocytosis
chemotaxis
inflammatory response
cytoskeleton organization
G-protein coupled receptor signaling pathway
cell-cell signaling
behavior
regulation of cell shape
response to toxic substance
positive regulation of gene expression
negative regulation of gene expression
T cell chemotaxis
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
calcium-mediated signaling
signaling
positive regulation of cell migration
negative regulation of bone mineralization
neutrophil chemotaxis
lipopolysaccharide-mediated signaling pathway
positive regulation of tumor necrosis factor production
eosinophil degranulation
protein kinase B signaling
positive regulation of neuron apoptotic process
positive regulation of GTPase activity
astrocyte cell migration
negative regulation by host of viral transcription
negative regulation of osteoclast differentiation
eosinophil chemotaxis
macrophage chemotaxis
lymphocyte chemotaxis
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of calcium-mediated signaling
positive chemotaxis
positive regulation of protein kinase B signaling
positive regulation of calcium ion transport
regulation of sensory perception of pain
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
response to cholesterol
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to organic cyclic compound
granulocyte chemotaxis
positive regulation of calcium ion import
positive regulation of natural killer cell chemotaxis
protein kinase activity
calcium-dependent protein kinase C activity
protein binding
chemokine activity
phospholipase activator activity
kinase activity
CCR1 chemokine receptor binding
CCR5 chemokine receptor binding
chemoattractant activity
identical protein binding
MAPK cascade
osteoblast differentiation
cell activation
monocyte chemotaxis
calcium ion transport
cellular calcium ion homeostasis
exocytosis
chemotaxis
inflammatory response
cytoskeleton organization
G-protein coupled receptor signaling pathway
cell-cell signaling
behavior
regulation of cell shape
response to toxic substance
positive regulation of gene expression
negative regulation of gene expression
T cell chemotaxis
release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
calcium-mediated signaling
signaling
positive regulation of cell migration
negative regulation of bone mineralization
neutrophil chemotaxis
lipopolysaccharide-mediated signaling pathway
positive regulation of tumor necrosis factor production
eosinophil degranulation
protein kinase B signaling
positive regulation of neuron apoptotic process
positive regulation of GTPase activity
astrocyte cell migration
negative regulation by host of viral transcription
negative regulation of osteoclast differentiation
eosinophil chemotaxis
macrophage chemotaxis
lymphocyte chemotaxis
positive regulation of interleukin-1 beta secretion
positive regulation of inflammatory response
positive regulation of calcium-mediated signaling
positive chemotaxis
positive regulation of protein kinase B signaling
positive regulation of calcium ion transport
regulation of sensory perception of pain
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
response to cholesterol
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to organic cyclic compound
granulocyte chemotaxis
positive regulation of calcium ion import
positive regulation of natural killer cell chemotaxis
protein kinase activity
calcium-dependent protein kinase C activity
protein binding
chemokine activity
phospholipase activator activity
kinase activity
CCR1 chemokine receptor binding
CCR5 chemokine receptor binding
chemoattractant activity
identical protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018376.hg.1 | ENST00000225245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018376.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018376.hg.1 | TC17001385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018377.hg.1 | ENST00000225245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018377.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17018377.hg.1 | TC17001385.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010459.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182126.hg.1 | TC1700010459.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | ENST00000613922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | ENST00000613928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | ENST00000614051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | NM_002983.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700182127.hg.1 | TC1700010459.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 131915 | - | - |

CCL3
Genomic Map