Assay Details
| Gene Symbol: ST20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100879.1 | NP_001094349.1 | 1 | 238 | 96 | |
| GenBank mRNA | AL109714.1 | - | 1 | 238 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15002781.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001100879.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
release of cytochrome c from mitochondria
mitochondrial fusion
intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of apoptotic process
transmembrane transport
extrinsic apoptotic signaling pathway in absence of ligand
channel activity
protein homodimerization activity
protein heterodimerization activity
release of cytochrome c from mitochondria
mitochondrial fusion
intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of apoptotic process
transmembrane transport
extrinsic apoptotic signaling pathway in absence of ligand
channel activity
protein homodimerization activity
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15016845.hg.1 | TC15002781.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15016845.hg.1 | ENST00000478497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15016845.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185606.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185606.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185606.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500185606.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500185606.hg.1 | TC1500010901.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000478497.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000485386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | ENST00000562759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100879.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001100880.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NM_001199757.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037652.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037653.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010901.hg.1 | NR_037653.1 | Refseq |

ST20
Genomic Map