Assay Details
Gene Symbol: KRTAP5-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004325.1 | NP_001004325.1 | 1 | 1043 | 104 | |
GenBank mRNA | AJ628245.1 | - | 1 | 1043 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001262.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07599366 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16409596 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07599358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27995374 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27995380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27995391 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07599368 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07599367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16409594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16409611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27995386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07599360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07599365 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27995378 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3364373 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0321 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45781 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3364373 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3364373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409591 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599357 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409591 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599357 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409596 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409594 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599346 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599344 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599341 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599371 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409594 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001004325_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7945649 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7945650 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7945651 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7937697 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7937696 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7945648 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1566218_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11735639_x_at | - | - |
Affymetrix Human Genome U219 Array | 11750368_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11735638_a_at | - | - |
Affymetrix Human Genome U219 Array | 11735638_PM_a_at | - | - |
Affymetrix Human Genome U219 Array | 11750369_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11735639_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11750369_x_at | - | - |
Affymetrix Human Genome U219 Array | 11750368_a_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11015972.hg.1 | ENST00000412090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11015972.hg.1 | NM_001004325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11015972.hg.1 | TC11001262.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000502328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000502328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000526239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000526239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000526239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000616086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000616086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000617152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000617152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | ENST00000617152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000398530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000502328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000502328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000526239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000526239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000526239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000616086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000616086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000617152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000617152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | ENST00000617152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011520.hg.1 | NM_001005405.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100173659.hg.1 | ENST00000412090.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100173659.hg.1 | NM_001004325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100173659.hg.1 | ENST00000412090.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100173659.hg.1 | NM_001004325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100173659.hg.1 | TC1100009746.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1566218_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1562681_at | - | - |
Human Genome U133 Plus 2.0 Array | 1562681_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1566218_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044745 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044745 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 228508 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1802360 | DA061271.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_26544-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 4780193 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_26544 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1802360 | AK055141.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1802360 | AJ628245.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1802360 | NM_001004325.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1802360 | DA061271.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1802360 | NR_021489.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1802360 | AK055141.1 | human_transcriptome |
