Assay Details
| Gene Symbol: KRTAP17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031964.1 | NP_114170.1 | 1 | 46 | 82 | |
| GenBank mRNA | AJ406952.1 | - | 1 | 46 | 82 | |
| BC069297.1 | - | 1 | 46 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001498.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ406952.1,BC069297.1,NM_031964.1 transcripts.
Set Membership: |
|

KRTAP17-1
Genomic Map