Assay Details
Gene Symbol: H2AFJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177925.3 | NP_808760.1 | 1 | 51 | 60 | |
NR_027716.2 | 1 | 51 | 60 | |||
GenBank mRNA | BM689354.1 | - | 1 | 55 | 60 | |
DA808847.1 | - | 1 | 51 | 60 | ||
DB084616.1 | - | 1 | 51 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000197.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_177925.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12002455.hg.1 | ENST00000544848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002455.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002455.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002455.hg.1 | TC12000197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12002456.hg.1 | ENST00000544848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002456.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002456.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002456.hg.1 | TC12000197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12002457.hg.1 | ENST00000228929.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002457.hg.1 | ENST00000544848.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12002457.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002457.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12002457.hg.1 | TC12000197.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148543.hg.1 | TC1200006946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148545.hg.1 | TC1200006946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148547.hg.1 | TC1200006946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000501744.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200006946.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148542.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148542.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148542.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148542.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148542.hg.1 | TC1200006946.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | ENST00000389078.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | ENST00000544848.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | NM_177925.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | NR_027716.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200148546.hg.1 | TC1200006946.hg.1 | Clariom D TC |
