Assay Details
| Gene Symbol: DUX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012148.2 | NP_036280.2 | 1 | 147 | 85 | |
| Gene Symbol: DUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012146.1 | NP_036278.1 | 1 | 178 | 85 | |
| GenBank mRNA | AJ001481.1 | - | 1 | 178 | 85 | |
| Gene Symbol: DUX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012149.2 | NP_036281.2 | 1 | 147 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P158257 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P158257 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 19879 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 691 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P158257 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P158257 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P158257 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P158257 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P158257 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8098742 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8098731 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8104123 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8098744 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8098730 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8104122 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8098740 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8098743 | - | - |
| Human Genome U133 Plus 2.0 Array | 208582_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208582_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208201_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208201_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208201_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208582_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208201_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208582_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 208582_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 208582_s_at | - | - |
| Human Genome U133A 2.0 Array | 208201_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208582_s_at | - | - |
| Human Genome U133A 2.0 Array | 208582_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 208201_at | - | - |
| Human Genome U133A 2.0 Array | 208201_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208201_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_012148_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_012149_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw555597 | - | - |
| Human Genome U95 Set | 31387_at | - | - |
| Human Genome U95 Set | 31387_PM_at | - | - |
| Human Genome U95 Set | 31387_at | - | - |
| Human Genome U95 Set | 31387_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208582_s_at1 | - | - |
| Human Genome U133 Set | 208582_PM_s_at | - | - |
| Human Genome U133 Set | 208582_s_at | - | - |
| Human Genome U133 Set | 208201_PM_at | - | - |
| Human Genome U133 Set | 208582_PM_s_at | - | - |
| Human Genome U133 Set | 208201_at | - | - |
| Human Genome U133 Set | 208582_s_at | - | - |
| Human Genome U133 Set | 208201_at | - | - |
| Human Genome U133 Set | 208201_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 375919 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_6912343-S | NM_012148.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11120737-S | NM_012146.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11120737-S | AJ001481.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11120737-S | NM_012148.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11120737-S | NM_012149.2 | human_transcriptome |
| Illumina human-6 v1.0 expression beadchip | 0000060458 | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006590670 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006590670 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0000060458 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2115813 | NM_012149.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2191067 | NM_012148.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115813 | NM_012146.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115813 | AJ001481.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115813 | NM_012148.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2115813 | NM_012149.2 | human_transcriptome |

DUX3
Genomic Map