Assay Details
Gene Symbol: ADORA2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028483.2 | 3 - 4 | 678 | 68 | ||
GenBank mRNA | BC045098.1 | - | 3 - 4 | 419 | 68 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BI829678.1 | - | 416 | Chr. - - [-] on Build | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SPECC1L-ADORA2A readthrough (NMD candidate) | SPECC1L-ADORA2A | -18.1kb | Anti-sense | Chr. 22 - 24270817 - 24442360 on NCBI Build GRCh38 | |
Downstream | beta-ureidopropionase 1 | UPB1 | BUP1 | 22.6kb | Anti-sense | Chr. 22 - 24495060 - 24528681 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000588.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010358.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22010358.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010358.hg.1 | TC22000588.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22010357.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22010357.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22010357.hg.1 | TC22000588.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | TC2200008218.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158892.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | TC2200008218.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158891.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000326341.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000412790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000427813.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | ENST00000543438.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008218.hg.1 | NR_028484.3 | Refseq |
