| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT HG-U133+ PM Array Plate |
1555865_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11000852.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11000852.hg.1 |
TC11000042.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11000853.hg.1 |
ENST00000530897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11000853.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11000853.hg.1 |
TC11000042.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006512.hg.1 |
ENST00000530897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006512.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100006512.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006512.hg.1 |
ENST00000530897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100006512.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100006512.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145415.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145415.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145415.hg.1 |
TC1100006512.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100145417.hg.1 |
ENST00000530897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145417.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145417.hg.1 |
ENST00000530897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100145417.hg.1 |
NR_029409.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100145417.hg.1 |
TC1100006512.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1555865_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1555865_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1555865_at9 |
- |
- |
| Human Genome U95 Set |
45851_at |
- |
- |
| Human Genome U95 Set |
45851_PM_at |
- |
- |