Assay Details
| Gene Symbol: MIR181A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029611.1 | 1 | 56 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR181A2 host gene | MIR181A2HG | Sense | Chr. 9 - 124658436 - 124698628 on NCBI Build GRCh38 | ||
| Downstream | microRNA 181b-2 | MIR181B2 |
MIRN181B2, mir-181b-2 |
1.2kb | Sense | Chr. 9 - 124693710 - 124693798 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0900008736.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000641.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302839 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302879 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957322 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302860 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302854 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302848 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16262477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957347 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302860 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302846 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302863 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957295 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302871 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302854 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957305 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957322 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302848 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302839 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27302879 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06957299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16262477 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8157800 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008736.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008736.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008736.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008736.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007362.hg.1 | TC09000641.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007362.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007362.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158245.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158245.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158245.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158245.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158245.hg.1 | TC0900008736.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008736.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008736.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008736.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008736.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158244.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158244.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158244.hg.1 | ENST00000384863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158244.hg.1 | NR_029611.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158244.hg.1 | TC0900008736.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309659 | NR_029611.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309659 | NR_029611.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309659 | NR_029611.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309659 | NR_029611.1 | human_transcriptome |

MIR181A2
Genomic Map