Assay Details
| Gene Symbol: MIR181B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029612.1 | 1 | 52 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR181A1 host gene | MIR181A1HG | Sense | Chr. 1 - 198808003 - 198937429 on NCBI Build GRCh38 | ||
| Downstream | microRNA 181a-1 | MIR181A1 |
MIR213, MIRN181A1, MIRN213, hsa-mir-181a-1, mir-181a-1, mir-213 |
62.0 | Sense | Chr. 1 - 198859044 - 198859153 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100016864.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003671.hg.1
Set Membership: |
|

MIR181B1
Genomic Map