Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
7924401 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017371.hg.1 |
ENST00000384858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017371.hg.1 |
NR_029628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058627.hg.1 |
TC01003836.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058627.hg.1 |
ENST00000384858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01058627.hg.1 |
NR_029628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017371.hg.1 |
ENST00000384858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017371.hg.1 |
NR_029628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231458.hg.1 |
ENST00000384858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231458.hg.1 |
NR_029628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231458.hg.1 |
ENST00000384858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100231458.hg.1 |
NR_029628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100231458.hg.1 |
TC0100017371.hg.1 |
Clariom D TC |