Assay Details
| Gene Symbol: MIR218-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029631.1 | 1 | 54 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SLIT2 intronic transcript 1 | SLIT2-IT1 | -133.4kb | Sense | Chr. 4 - 20392189 - 20394856 on NCBI Build GRCh38 | |
| Downstream | PARK2 coregulated like | PACRGL | C4orf28 | 167.9kb | Sense | Chr. 4 - 20696282 - 20757766 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0400007020.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000160.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15550727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119987 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120018 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119991 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119970 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119995 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119967 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967137 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15550727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967067 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967088 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120024 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967141 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119973 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119970 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119976 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119996 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119995 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120004 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119967 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119982 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119989 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120015 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120014 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119983 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967071 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967095 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119987 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967082 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967127 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119991 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967074 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120000 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119985 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02967137 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120018 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23119994 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23120008 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8094340 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002511.hg.1 | TC04000160.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002511.hg.1 | ENST00000384999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04002511.hg.1 | NR_029631.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007020.hg.1 | ENST00000384999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007020.hg.1 | NR_029631.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148186.hg.1 | ENST00000384999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148186.hg.1 | NR_029631.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148186.hg.1 | ENST00000384999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400148186.hg.1 | NR_029631.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400148186.hg.1 | TC0400007020.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400007020.hg.1 | ENST00000384999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007020.hg.1 | NR_029631.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3308565 | NR_029631.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3308565 | NR_029631.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308565 | NR_029631.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308565 | NR_029631.1 | human_transcriptome |

MIR218-1
Genomic Map