Assay Details
| Gene Symbol: MIR153-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029688.1 | 1 | 46 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, receptor type N |
PTPRN |
IA-2, IA-2/PTP, IA2, ICA512, R-PTP-N |
Sense | Chr. 2 - 219289623 - 219309573 on NCBI Build GRCh38 | |
| Downstream | regulated endocrine specific protein 18 | RESP18 | 33.2kb | Sense | Chr. 2 - 219327409 - 219336656 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200015774.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002795.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15384331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15384335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901237 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901191 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026738 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901185 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901172 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901215 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901196 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901174 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901217 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901188 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15384335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901226 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026743 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901176 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901205 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901236 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901208 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901211 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901181 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901195 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15384331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901199 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901210 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901190 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901187 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901177 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01901202 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22026732 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 8059211 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045186.hg.1 | TC02002795.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045186.hg.1 | ENST00000384914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02045186.hg.1 | NR_029688.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015774.hg.1 | ENST00000384914.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015774.hg.1 | NR_029688.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200216131.hg.1 | ENST00000384914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200216131.hg.1 | NR_029688.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200216131.hg.1 | ENST00000384914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200216131.hg.1 | NR_029688.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200216131.hg.1 | TC0200015774.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015774.hg.1 | ENST00000384914.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015774.hg.1 | NR_029688.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309684 | NR_029688.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309684 | NR_029688.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309684 | NR_029688.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309684 | NR_029688.1 | human_transcriptome |

MIR153-1
Genomic Map