Assay Details
| Gene Symbol: MIR365B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029856.1 | 1 | 50 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4725 | MIR4725 | mir-4725 | -53.0 | Sense | Chr. 17 - 31575269 - 31575358 on NCBI Build GRCh38 |
| Downstream | coordinator of PRMT5 and differentiation stimulator | COPRS |
C17orf79, COPR5, HSA272196, TTP1 |
276.3kb | Anti-sense | Chr. 17 - 31851865 - 31859307 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1700007487.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000366.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16969452 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850160 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284100 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284096 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16969446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850132 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284103 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284097 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16969447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850129 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850120 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284098 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284121 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850118 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850119 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284138 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16969446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850101 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284139 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850130 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850108 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850104 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284111 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850160 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16969447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284103 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284096 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284122 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284085 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284117 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850136 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284126 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850147 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284100 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16969452 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850102 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10284097 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850155 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30850132 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17004634.hg.1 | ENST00000362317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004634.hg.1 | NR_029856.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004634.hg.1 | TC17000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007487.hg.1 | ENST00000362317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007487.hg.1 | ENST00000412403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007487.hg.1 | ENST00000412403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007487.hg.1 | ENST00000412403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007487.hg.1 | NR_029856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153781.hg.1 | ENST00000362317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153781.hg.1 | NR_029856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153781.hg.1 | ENST00000362317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153781.hg.1 | NR_029856.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153781.hg.1 | TC1700007487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007487.hg.1 | ENST00000362317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007487.hg.1 | ENST00000412403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007487.hg.1 | ENST00000412403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007487.hg.1 | ENST00000412403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007487.hg.1 | NR_029856.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310366 | NR_029856.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310366 | NR_029856.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310366 | NR_029856.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310366 | NR_029856.1 | human_transcriptome |

MIR365B
Genomic Map