Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.1 ST Array |
7896863 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018154.hg.1 |
ENST00000362106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018154.hg.1 |
NR_029957.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000307.hg.1 |
TC01000029.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000307.hg.1 |
ENST00000362106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000307.hg.1 |
NR_029957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236672.hg.1 |
ENST00000362106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236672.hg.1 |
NR_029957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236672.hg.1 |
ENST00000362106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100236672.hg.1 |
NR_029957.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100236672.hg.1 |
TC0100018154.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018154.hg.1 |
ENST00000362106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018154.hg.1 |
NR_029957.1 |
Refseq |