Assay Details
Gene Symbol: MIR490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030165.1 | 1 | 68 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine zipper protein 6 | LUZP6 |
MPD6, MTPNUT |
-925.7kb | Anti-sense | Chr. 7 - 135926755 - 135977456 on NCBI Build GRCh38 |
Downstream | pleiotrophin | PTN |
HARP, HB-GAM, HBBM, HBGF-8, HBGF8, HBNF, HBNF-1, NEGF1, OSF-1 |
324.0kb | Anti-sense | Chr. 7 - 137227341 - 137343865 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700009278.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000853.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070654 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812496 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070646 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812467 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812508 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812478 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812517 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070661 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070656 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812465 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812453 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812496 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812465 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070650 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812517 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812467 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812478 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812508 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070654 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070638 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070637 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812501 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070652 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070656 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070631 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070646 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070663 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070627 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812453 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070661 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070684 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26070648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05812495 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07013422.hg.1 | TC07000853.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07013422.hg.1 | ENST00000384865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07013422.hg.1 | NR_030165.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700009278.hg.1 | ENST00000384865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009278.hg.1 | NR_030165.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700009278.hg.1 | ENST00000384865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009278.hg.1 | NR_030165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700166008.hg.1 | ENST00000384865.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700166008.hg.1 | NR_030165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700166008.hg.1 | ENST00000384865.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700166008.hg.1 | NR_030165.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700166008.hg.1 | TC0700009278.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3311080 | NR_030165.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3311080 | NR_030165.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3311080 | NR_030165.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3311080 | NR_030165.1 | human_transcriptome |
