Assay Details
| Gene Symbol: MIR487B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030267.1 | 1 | 42 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 381 | MIR381 |
MIRN381, hsa-mir-381, mir-381 |
-461.0 | Sense | Chr. 14 - 101045920 - 101045994 on NCBI Build GRCh38 |
| Downstream | microRNA 539 | MIR539 |
MIRN539, hsa-mir-539, mir-539 |
783.0 | Sense | Chr. 14 - 101047321 - 101047398 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1400008289.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000743.hg.1
Set Membership: |
|

MIR487B
Genomic Map