Assay Details
Gene Symbol: MIR592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030323.1 | 1 | 55 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928283 | LOC101928283 | -1.7mb | Anti-sense | Chr. 7 - 125184569 - 125379321 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101928333 | LOC101928333 | 156.9kb | Anti-sense | Chr. 7 - 127215127 - 127229921 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012497.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001834.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023566 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023552 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023546 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023532 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023566 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766659 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023534 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023570 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766641 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023532 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766672 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023546 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05766642 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023552 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26023562 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07028420.hg.1 | TC07001834.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07028420.hg.1 | ENST00000384959.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07028420.hg.1 | NR_030323.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700012497.hg.1 | ENST00000384959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012497.hg.1 | NR_030323.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700190051.hg.1 | ENST00000384959.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700190051.hg.1 | NR_030323.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700190051.hg.1 | ENST00000384959.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700190051.hg.1 | NR_030323.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700190051.hg.1 | TC0700012497.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012497.hg.1 | ENST00000384959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700012497.hg.1 | NR_030323.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310341 | NR_030323.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310341 | NR_030323.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310341 | NR_030323.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310341 | NR_030323.1 | human_transcriptome |
