Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011826.hg.1 |
TC01000735.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01011826.hg.1 |
NR_036060.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008633.hg.1 |
ENST00000584034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008633.hg.1 |
ENST00000584034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008633.hg.1 |
NR_036060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008633.hg.1 |
ENST00000584034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008633.hg.1 |
ENST00000584034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008633.hg.1 |
NR_036060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161425.hg.1 |
ENST00000584034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161425.hg.1 |
NR_036060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161425.hg.1 |
ENST00000584034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100161425.hg.1 |
NR_036060.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100161425.hg.1 |
TC0100008633.hg.1 |
Clariom D TC |