Assay Details
| Gene Symbol: MIR3136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036087.1 | 1 | 40 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EGF domain specific O-linked N-acetylglucosamine transferase | EOGT |
AER61, AOS4, C3orf64, EOGT1 |
-35.0kb | Sense | Chr. 3 - 68975212 - 69013961 on NCBI Build GRCh38 |
| Downstream | ubiquitin like modifier activating enzyme 3 | UBA3 |
NAE2, UBE1C, hUBA3 |
5.7kb | Sense | Chr. 3 - 69054730 - 69080373 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0300011451.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001537.hg.1
Set Membership: |
|

MIR3136
Genomic Map