Assay Details
| Gene Symbol: MIR3141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036094.1 | 1 | 34 | 52 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heart and neural crest derivatives expressed 1 | HAND1 |
Hxt, Thing1, bHLHa27, eHand |
-117.7kb | Sense | Chr. 5 - 154474972 - 154478264 on NCBI Build GRCh38 |
| Downstream | La ribonucleoprotein domain family member 1 | LARP1 | LARP | 86.6kb | Anti-sense | Chr. 5 - 154682663 - 154817607 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0500012556.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001972.hg.1
Set Membership: |
|

MIR3141
Genomic Map