Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019669.hg.1 |
NR_036113.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019669.hg.1 |
TC10001554.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011510.hg.1 |
ENST00000578688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011510.hg.1 |
NR_036113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011510.hg.1 |
ENST00000578688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011510.hg.1 |
NR_036113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178202.hg.1 |
ENST00000578688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178202.hg.1 |
NR_036113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178202.hg.1 |
ENST00000578688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178202.hg.1 |
NR_036113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178202.hg.1 |
TC1000011510.hg.1 |
Clariom D TC |