Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009156.hg.1 |
NR_036147.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009156.hg.1 |
TC16000649.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008626.hg.1 |
ENST00000580839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008626.hg.1 |
NR_036147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164341.hg.1 |
ENST00000580839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164341.hg.1 |
NR_036147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164341.hg.1 |
ENST00000580839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600164341.hg.1 |
NR_036147.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600164341.hg.1 |
TC1600008626.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008626.hg.1 |
ENST00000580839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008626.hg.1 |
NR_036147.1 |
Refseq |