Assay Details
| Gene Symbol: MIR3186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036152.1 | 1 | 47 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4740 | MIR4740 | -50.3kb | Sense | Chr. 17 - 81400716 - 81400778 on NCBI Build GRCh38 | |
| Downstream | actin gamma 1 | ACTG1 |
ACT, ACTG, BRWS2, DFNA20, DFNA26, HEL-176 |
58.8kb | Sense | Chr. 17 - 81509971 - 81512866 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1700012048.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001951.hg.1
Set Membership: |
|

MIR3186
Genomic Map