Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024325.hg.1 |
NR_036189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024325.hg.1 |
TC12001854.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011637.hg.1 |
ENST00000581299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011637.hg.1 |
NR_036189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190107.hg.1 |
ENST00000581299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190107.hg.1 |
NR_036189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190107.hg.1 |
ENST00000581299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200190107.hg.1 |
NR_036189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200190107.hg.1 |
TC1200011637.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011637.hg.1 |
ENST00000581299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011637.hg.1 |
NR_036189.1 |
Refseq |