Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007609.hg.1 |
ENST00000580794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007609.hg.1 |
NR_036196.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004979.hg.1 |
NR_036196.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15004979.hg.1 |
TC15000610.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500153907.hg.1 |
ENST00000580794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153907.hg.1 |
NR_036196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153907.hg.1 |
ENST00000580794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500153907.hg.1 |
NR_036196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500153907.hg.1 |
TC1500007609.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007609.hg.1 |
ENST00000580794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007609.hg.1 |
NR_036196.1 |
Refseq |