Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025894.hg.1 |
NR_036200.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025894.hg.1 |
TC17001910.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011859.hg.1 |
ENST00000579394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011859.hg.1 |
NR_036200.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011859.hg.1 |
ENST00000579394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011859.hg.1 |
NR_036200.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700196778.hg.1 |
ENST00000579394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196778.hg.1 |
NR_036200.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700196778.hg.1 |
ENST00000579394.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700196778.hg.1 |
NR_036200.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700196778.hg.1 |
TC1700011859.hg.1 |
Clariom D TC |