Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035472.hg.1 |
TC02002185.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035472.hg.1 |
NR_036223.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013827.hg.1 |
ENST00000582152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013827.hg.1 |
NR_036223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013827.hg.1 |
ENST00000582152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013827.hg.1 |
NR_036223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201201.hg.1 |
ENST00000582152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201201.hg.1 |
NR_036223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201201.hg.1 |
ENST00000582152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200201201.hg.1 |
NR_036223.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200201201.hg.1 |
TC0200013827.hg.1 |
Clariom D TC |