Assay Details
Gene Symbol: LOC100128361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036505.1 | 1 | 154 | 92 | ||
GenBank mRNA | AK094300.1 | - | 1 | 154 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centromere protein P | CENPP | CENP-P | -2.9kb | Sense | Chr. 9 - 92325457 - 92615164 on NCBI Build GRCh38 |
Downstream | BICD cargo adaptor 2 | BICD2 |
SMALED2, bA526D8.1 |
90.8kb | Anti-sense | Chr. 9 - 92711363 - 92764841 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000436.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P93996 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P93991 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09004617.hg.1 | TC09000436.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09004617.hg.1 | ENST00000375587.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004617.hg.1 | NR_036505.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007953.hg.1 | NR_036505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153536.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153536.hg.1 | NR_036505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153536.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153536.hg.1 | NR_036505.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153536.hg.1 | TC0900007953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375576.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375579.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000375587.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | ENST00000618653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001012267.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286969.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NM_001286971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007953.hg.1 | NR_036505.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 242207_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 242207_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 242207_at1 | - | - |
Human Genome U133 Set | 242207_at | - | - |
Human Genome U133 Set | 242207_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 230676 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3272826 | - | - |
