Assay Details
Gene Symbol: LOC100131347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036551.1 | 2 - 3 | 263 | 86 | ||
GenBank mRNA | AK095115.1 | - | 2 - 3 | 263 | 86 | |
BC072408.1 | - | 2 - 3 | 216 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | plexin domain containing 1 | PLXDC1 |
TEM3, TEM7 |
Anti-sense | Chr. 17 - 39063303 - 39151649 on NCBI Build GRCh38 | |
Downstream | ADP ribosylation factor like GTPase 5C | ARL5C | ARL12 | 84.3kb | Anti-sense | Chr. 17 - 39156894 - 39166161 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000472.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30883211 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 908406 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 160341 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3549091 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24747 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3549091 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3549091 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20641599 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20641601 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20641603 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20641601 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1564050_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000410562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000577328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000577328.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000584586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | ENST00000584586.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005528.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005528.hg.1 | TC17000472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17005529.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005529.hg.1 | TC17000472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17005530.hg.1 | ENST00000410562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17005530.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005530.hg.1 | TC17000472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17005527.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005527.hg.1 | TC17000472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17005531.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17005531.hg.1 | TC17000472.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000593041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | ENST00000593041.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155809.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155809.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155809.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155809.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155809.hg.1 | TC1700007732.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | ENST00000410562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | ENST00000410562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155812.hg.1 | TC1700007732.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700155813.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155813.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155813.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155813.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155813.hg.1 | TC1700007732.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700155810.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155810.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155810.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155810.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155810.hg.1 | TC1700007732.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000410562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000577328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000577328.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000578423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000584586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | ENST00000584586.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007732.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000592982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000593041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | ENST00000593041.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007773.hg.1 | NR_026887.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155811.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155811.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155811.hg.1 | ENST00000583447.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700155811.hg.1 | NR_036551.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700155811.hg.1 | TC1700007732.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1564050_at | - | - |
Human Genome U133 Plus 2.0 Array | 240963_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 1564050_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 240963_x_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240963_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1564050_at10 | - | - |
Human Genome U95 Set | 65055_at | - | - |
Human Genome U95 Set | 65055_PM_at | - | - |
Human Genome U133 Set | 240963_PM_x_at | - | - |
Human Genome U133 Set | 240963_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009456 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009456 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3208344 | - | - |
