Assay Details
Gene Symbol: DPY19L2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ287933.1 | - | 2 - 3 | 216 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc and ring finger 2 pseudogene 2 | ZNRF2P2 | -1.5kb | Sense | Chr. 7 - 29684772 - 29685821 on NCBI Build GRCh38 | |
Downstream | probable C-mannosyltransferase DPY19L2 | LOC101928208 | 29.5kb | Anti-sense | Chr. 7 - 29720476 - 29742396 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
