Assay Details
Gene Symbol: DPY19L2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ287933.1 | - | 5 - 6 | 479 | 186 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | probable C-mannosyltransferase DPY19L2 | LOC101928208 | 10.4kb | Sense | Chr. 7 - 29720476 - 29742396 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
