Assay Details
| Gene Symbol: SMIM10L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271592.1 | NP_001258521.1 | 1 | 47 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PRH1-PRR4 readthrough | PRH1-PRR4 | PRH1 | Anti-sense | Chr. 12 - 10845849 - 11171625 on NCBI Build GRCh38 | |
| Downstream | taste 2 receptor member 42 | TAS2R42 |
T2R24, T2R42, T2R55, TAS2R55 |
10.0kb | Anti-sense | Chr. 12 - 11185993 - 11186937 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271592.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SMIM10L1
Genomic Map