Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031243.hg.1 |
TC07002039.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031243.hg.1 |
NR_037468.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013077.hg.1 |
ENST00000579424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013077.hg.1 |
NR_037468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194401.hg.1 |
ENST00000579424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194401.hg.1 |
NR_037468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194401.hg.1 |
ENST00000579424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194401.hg.1 |
NR_037468.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194401.hg.1 |
TC0700013077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013077.hg.1 |
ENST00000579424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013077.hg.1 |
NR_037468.1 |
Refseq |