Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006425.hg.1 |
NR_037499.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006425.hg.1 |
TC16000462.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007945.hg.1 |
ENST00000583472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007945.hg.1 |
NR_037499.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007945.hg.1 |
ENST00000583472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007945.hg.1 |
NR_037499.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157942.hg.1 |
ENST00000583472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157942.hg.1 |
NR_037499.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157942.hg.1 |
ENST00000583472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600157942.hg.1 |
NR_037499.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600157942.hg.1 |
TC1600007945.hg.1 |
Clariom D TC |