Assay Details
Gene Symbol: LHX4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037642.1 | 4 - 5 | 343 | 109 | ||
GenBank mRNA | AK096250.1 | - | 4 - 5 | 343 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized protein FLJ23867 | FLJ23867 | -68.9kb | Anti-sense | Chr. 1 - 180198009 - 180200725 on NCBI Build GRCh38 | |
Downstream | acyl-CoA binding domain containing 6 | ACBD6 | 14.7kb | Sense | Chr. 1 - 180269663 - 180502967 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01006384.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 8x60K | 35665 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 923799 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 389820 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P519638 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 2463 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P519638 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P519638 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00202930 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00202930 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01055339.hg.1 | TC01006384.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01055339.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01055339.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244230.hg.1 | TC0100018539.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244229.hg.1 | TC0100018539.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000440959.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | ENST00000622400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018539.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244227.hg.1 | TC0100018539.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | ENST00000415414.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | NR_037642.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100244228.hg.1 | TC0100018539.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008577 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008577 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 232623 | - | - |
