Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
ENST00000404721.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009276.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010955.hg.1 |
TC06000936.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010955.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06010955.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
ENST00000404721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009276.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
ENST00000404721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
ENST00000404721.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163677.hg.1 |
TC0600009276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163678.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163678.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163678.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163678.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163678.hg.1 |
TC0600009276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600163676.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163676.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163676.hg.1 |
ENST00000429807.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600163676.hg.1 |
NR_037662.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600163676.hg.1 |
TC0600009276.hg.1 |
Clariom D TC |