Assay Details
| Gene Symbol: CES5AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037839.1 | 1 - 2 | 159 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
BCR, RhoGEF and GTPase activating protein |
BCR |
ALL, BCR1, CML, D22S11, D22S662, PHL |
-58.8kb | Anti-sense | Chr. 22 - 23180365 - 23318037 on NCBI Build GRCh38 |
| Downstream | zinc finger DHHC-type containing 8 pseudogene 1 | ZDHHC8P1 | ZDHHC8P | 8.5kb | Sense | Chr. 22 - 23390605 - 23402612 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000558.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010071.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010071.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010071.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010073.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010073.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010073.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010072.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010072.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010072.hg.1 | TC22000558.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158365.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000422151.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008164.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000400418.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000424029.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | ENST00000431237.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158368.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158374.hg.1 | TC2200008164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158374.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158374.hg.1 | NR_037839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158374.hg.1 | ENST00000415114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158374.hg.1 | NR_037839.1 | Refseq |

CES5AP1
Genomic Map