Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037921.1 | 3 - 4 | 463 | 61 | ||
| GenBank mRNA | AK296897.1 | - | 3 - 4 | 428 | 61 | |
| EF467324.1 | - | 2 - 3 | 291 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adaptor related protein complex 5 sigma 1 subunit | AP5S1 | C20orf29 | -32.3kb | Sense | Chr. 20 - 3820524 - 3825307 on NCBI Build GRCh38 |
| Downstream | pantothenate kinase 2 | PANK2 |
C20orf48, HARP, HSS, NBIA1, PKAN |
26.4kb | Sense | Chr. 20 - 3888839 - 3929882 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000043.hg.1
Set Membership: |
|

MAVS
Genomic Map