Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206491.1 | NP_001193420.1 | 2 - 3 | 290 | 82 | |
| GenBank mRNA | AK291785.1 | - | 2 - 3 | 249 | 82 | |
| BX649084.1 | - | 2 - 3 | 251 | 82 | ||
| EF467323.1 | - | 1 - 2 | 118 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adaptor related protein complex 5 sigma 1 subunit | AP5S1 | C20orf29 | -29.3kb | Sense | Chr. 20 - 3820524 - 3825307 on NCBI Build GRCh38 |
| Downstream | pantothenate kinase 2 | PANK2 |
C20orf48, HARP, HSS, NBIA1, PKAN |
27.3kb | Sense | Chr. 20 - 3888839 - 3929882 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK291785.1, NM_001206491.1 transcripts.
Set Membership: |
|

MAVS
Genomic Map