Assay Details
Gene Symbol: DYX1C1-CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037923.1 | 8 - 9 | 1403 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 628 | MIR628 |
MIRN628, hsa-mir-628, mir-628 |
-16.3kb | Sense | Chr. 15 - 55372940 - 55373034 on NCBI Build GRCh38 |
Downstream | pygopus family PHD finger 1 | PYGO1 | 106.3kb | Sense | Chr. 15 - 55538884 - 55588852 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184649.hg.1 | TC1500010868.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184648.hg.1 | TC1500010868.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500184650.hg.1 | TC1500010868.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000565113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | ENST00000568310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010868.hg.1 | NR_037923.1 | Refseq |
