Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
ENST00000243997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
ENST00000395659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
ENST00000395663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
NM_006886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
NR_037929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
NR_037930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
TC2000009648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
ENST00000243997.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
ENST00000395659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
ENST00000395663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
NM_006886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
NR_037929.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000167315.hg.1 |
NR_037930.1 |
Refseq |