Assay Details
| Gene Symbol: ANAPC1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037931.1 | 14 - 15 | 1684 | 71 | ||
| GenBank mRNA | AK310642.1 | - | 14 - 15 | 1684 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CD8b molecule | CD8B |
CD8B1, LEU2, LY3, LYT3, P37 |
-31.4kb | Anti-sense | Chr. 2 - 86815337 - 86861924 on NCBI Build GRCh38 |
| Downstream | RANBP2-like and GRIP domain containing 1 | RGPD1 |
RGP1, RGPD2, RanBP2L2 |
18.7kb | Sense | Chr. 2 - 86913575 - 87013976 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000527.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21419146 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21419223 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P21419206 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28284 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3279851 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3363655 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8043323 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8043322 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008267.hg.1 | TC02000527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008267.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008266.hg.1 | TC02000527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008266.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008265.hg.1 | TC02000527.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02008265.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222539.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200222539.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222539.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200222539.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222539.hg.1 | TC0200016483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200222538.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200222538.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222538.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200222538.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222538.hg.1 | TC0200016483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200222540.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200222540.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222540.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200222540.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200222540.hg.1 | TC0200016483.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | ENST00000616781.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200016483.hg.1 | NR_037931.1 | Refseq |
| Human Genome U95 Set | 52704_at | - | - |
| Human Genome U95 Set | 52704_PM_at | - | - |
| Human Genome U95 Set | 58914_PM_at | - | - |
| Human Genome U95 Set | 58914_at | - | - |
| Human Genome U133 Set | 229267_PM_at | - | - |
| Human Genome U133 Set | 229267_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015926 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015927 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015927 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A015926 | - | - |

ANAPC1P1
Genomic Map