Assay Details
Gene Symbol: STX16-NPEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037945.1 | 9 - 10 | 1824 | 98 | ||
GenBank mRNA | AB209296.1 | - | 9 - 10 | 1643 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | aminopeptidase-like 1 | NPEPL1 | bA261P9.2 | Sense | Chr. 20 - 58689131 - 58715844 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC105372695 | LOC105372695 | 81.8kb | Sense | Chr. 20 - 58773425 - 58774811 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000413559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000413559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000413559.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170261.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170261.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000170261.hg.1 | TC2000009947.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000170261.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000170261.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000413559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000413559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000413559.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | ENST00000530122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009947.hg.1 | NR_037945.1 | Refseq |
