Assay Details
| Gene Symbol: SBF1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027765.2 | 1 | 2848 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | XK related 4 | XKR4 | XRG4 | Anti-sense | Chr. 8 - 55102457 - 55526155 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105375843 | LOC105375843 | 59.1kb | Sense | Chr. 8 - 55517187 - 55520978 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001222.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416150 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5206 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5205 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53908 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416150 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3416150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417021 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133479 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133479 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417017 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133473 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417005 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417011 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06133471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26417021 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133448 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16096472 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133463 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133453 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16096468 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16096477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133450 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133456 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133451 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16096466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133487 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16096487 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16096461 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133443 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133458 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06133454 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08015853.hg.1 | TC08001222.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015853.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171260.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171260.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171260.hg.1 | TC0800010484.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800171261.hg.1 | ENST00000444527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171261.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171261.hg.1 | ENST00000444527.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171261.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171261.hg.1 | TC0800010484.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800171262.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171262.hg.1 | NR_027765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171262.hg.1 | TC0800010484.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035779 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035778 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035779 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035778 | - | - |

SBF1P1
Genomic Map