Assay Details
| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028302.1 | 1 - 2 | 393 | 88 | ||
| GenBank mRNA | AK309100.1 | - | 1 - 2 | 393 | 88 | |
| DA339707.1 | - | 1 - 2 | 393 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000538.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03009882.hg.1 | TC03000538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009882.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009882.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009882.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009883.hg.1 | TC03000538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009883.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009883.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009883.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009889.hg.1 | TC03000538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009889.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009884.hg.1 | TC03000538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009884.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009884.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009884.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158664.hg.1 | TC0300008264.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158661.hg.1 | TC0300008264.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158665.hg.1 | TC0300008264.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000466734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000473550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | ENST00000495228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028301.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008264.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | ENST00000463143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | ENST00000490441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | NR_028302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158684.hg.1 | TC0300008264.hg.1 | Clariom D TC |

DUBR
Genomic Map