Assay Details
Gene Symbol: ACVR2B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028389.1 | 1 - 2 | 321 | 87 | ||
GenBank mRNA | AK025619.1 | - | 1 - 2 | 321 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | xylulokinase | XYLB | Anti-sense | Chr. 3 - 38346760 - 38453041 on NCBI Build GRCh38 | ||
Downstream |
endo/exonuclease (5'-3'), endonuclease G-like |
EXOG |
ENDOGL1, ENDOGL2, ENGL, ENGL-a, ENGL-b, ENGLA, ENGLB |
41.3kb | Anti-sense | Chr. 3 - 38496127 - 38526305 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001295.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 781508 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3244478 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 31604 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3244478 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3244478 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313732 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180126 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180175 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180120 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180178 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180173 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180171 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180176 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313733 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180162 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313769 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313734 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180198 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180168 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180145 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180142 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180199 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180185 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180209 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180128 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180181 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180209 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313819 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313774 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180179 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180178 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313734 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313758 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180120 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313784 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180142 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313733 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313731 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180162 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180176 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180150 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180185 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180126 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313790 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313769 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180145 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180180 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180177 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180146 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313732 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180168 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180175 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180166 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180199 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313815 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180198 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180194 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180128 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180173 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180171 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22313821 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02180190 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00686442 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 215590_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR03022249.hg.1 | TC03001295.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03022249.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03022249.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010754.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010754.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010754.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010754.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03022248.hg.1 | TC03001295.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03022248.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03022248.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300178289.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300178289.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300178289.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300178289.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300178289.hg.1 | TC0300010754.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300010754.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010754.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010754.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010754.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300178291.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300178291.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300178291.hg.1 | ENST00000441531.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300178291.hg.1 | NR_028389.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300178291.hg.1 | TC0300010754.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 215590_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 215590_PM_x_at | - | - |
Human Genome U133A 2.0 Array | 215590_x_at | - | - |
Human Genome U133A 2.0 Array | 215590_PM_x_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw575651 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 215590_x_at5 | - | - |
Human Genome U133 Set | 215590_PM_x_at | - | - |
Human Genome U133 Set | 215590_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020893 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020893 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3179067 | AK025619.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3179067 | NR_028389.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3251776 | AK025619.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3251776 | NR_028389.1 | human_transcriptome |
